missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidone hydrochloride monohydrate, 98%

Product Code. p-7021407
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11414744

Brand: Thermo Scientific Alfa Aesar A13726.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 40064-34-4 | C5H9NO·HCl·H2O

4-Piperidone hydrochloride monohydrate is used in the generation of compound libraries based on sub-reactions of an Ugi multicomponent reaction (MCR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Piperidone hydrochloride monohydrate is used in the generation of compound libraries based on sub-reactions of an Ugi multicomponent reaction (MCR).

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40064-34-4
Molecular Formula C5H9NO·HCl·H2O
MDL Number MFCD00012776
Synonym piperidine-4,4-diol hydrate hydrochloride

Specifications

Melting Point 94°C to 100°C
Quantity 10 g
Beilstein 3916558
Solubility Information Soluble in water.
Formula Weight 153.61 (135.59 Anhydrous)
Percent Purity 98%
Chemical Name or Material 4-Piperidone hydrochloride monohydrate
compliance-icons
Product Identifier
  • 4-Piperidone hydrochloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Piperonylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.