missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidone ethylene ketal, 98+%, Thermo Scientific Chemicals

Product Code. 11409613
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

Solubility
Soluble in water (Partly miscible).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177-11-7
Molecular Formula C7H14NO2
Molecular Weight (g/mol) 144.19
MDL Number MFCD00005976
InChI Key KPKNTUUIEVXMOH-UHFFFAOYSA-O
Synonym 1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
PubChem CID 67435
SMILES C1COC2(CC[NH2+]CC2)O1

Specifications

Density 1.117
Boiling Point 99°C to 101°C (14 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.4819
Quantity 10 g
Beilstein 506799
Sensitivity Air Sensitive
Solubility Information Soluble in water (Partly miscible).
IUPAC Name 1,4-dioxa-8-azaspiro[4.5]decan-8-ium
Formula Weight 143.19
Percent Purity ≥98%
Chemical Name or Material 4-Piperidone ethylene ketal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'