missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidinopiperidine, 95%, Tech., Thermo Scientific™

Product Code. 10133111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133111 50 g Glass bottle 50g
10531421 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133111 Supplier Thermo Scientific Acros Supplier No. 165450500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4897-50-1
Molecular Formula C10H20N2
Molecular Weight (g/mol) 168.28
MDL Number MFCD00006475
InChI Key QDVBKXJMLILLLB-UHFFFAOYSA-N
Synonym 4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine
PubChem CID 78607
ChEBI CHEBI:40117
IUPAC Name 1-piperidin-4-ylpiperidine
SMILES C1CCN(CC1)C2CCNCC2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 62.0°C to 67.0°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 50 g
Formula Weight 168.28
Percent Purity 95%
Grade Technical
Physical Form Crystalline Solid
Chemical Name or Material 4-Piperidinopiperidine, tech., 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Piperidinopiperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.