missing translation for 'onlineSavingsMsg'
Learn More

(4-Piperidinophenyl)methylamine, 97%, Thermo Scientific™

Product Code. p-4510510
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10286242

Brand: Thermo Scientific Maybridge CC33913DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 214759-73-6
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD06659080
InChI Key WROBJEHXMLQDQP-UHFFFAOYSA-N
Synonym 4-piperidinophenyl methylamine, 4-piperidin-1-yl phenyl methanamine, 4-piperidinobenzylamine, 4-piperidin-1-yl-benzylamine, 1-4-piperidin-1-yl phenyl methanamine, 4-piperidin-1yl benzylamine, 4-piperidin-1ylphenyl methylamine, 4-piperidylphenyl methylamine, 4-1-piperidyl phenyl methanamine
PubChem CID 2795551
IUPAC Name (4-piperidin-1-ylphenyl)methanamine
SMILES NCC1=CC=C(C=C1)N1CCCCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material (4-Piperidinophenyl)methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.