missing translation for 'onlineSavingsMsg'
Learn More

4-(piperidinomethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10026202
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10026202 1 g Amber Glass Bottle 1g
10760564 250 mg Amber Glass Bottle 250mg
10516524 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10026202 Supplier Thermo Scientific Maybridge Supplier No. CC45501DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 159691-33-5
Molecular Formula C13H17NO2
Molecular Weight (g/mol) 219.284
InChI Key RNCCMTVMMFUIKR-UHFFFAOYSA-N
Synonym 4-piperidin-1-ylmethyl benzoic acid, 4-piperidin-1-ylmethyl-benzoic acid, benzoic acid,4-1-piperidinylmethyl, 4-piperidylmethyl benzoic acid, 4-piperidinomethyl benzoic acid, 4-piperidin-1-yl methyl benzoic acid
PubChem CID 1092975
IUPAC Name 4-(piperidin-1-ylmethyl)benzoic acid
SMILES C1CCN(CC1)CC2=CC=C(C=C2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material 4-(Piperidinomethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.