missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidinobenzoic acid, 97%, Thermo Scientific™

Product Code. 10026112
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10026112 1 g 1g
10138142 250 mg 250mg
10168794 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10026112 Supplier Thermo Scientific Maybridge Supplier No. CC33901DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22090-24-0
Molecular Formula C12H14NO2
Molecular Weight (g/mol) 204.25
MDL Number MFCD00587602
InChI Key DGJNUESQBVPXAY-UHFFFAOYSA-M
PubChem CID 764599
IUPAC Name 4-piperidin-1-ylbenzoic acid
SMILES [O-]C(=O)C1=CC=C(C=C1)N1CCCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 4-piperidin-1-yl-benzoic acid, 4-piperidin-1-yl benzoic acid, 4-piperidinobenzoic acid, 4-piperidinobenzenecarboxylic acid, benzoic acid, 4-1-piperidinyl, 1-4-carboxyphenyl piperidine, 4-1-piperidinyl benzoic acid, benzoic acid,4-1-piperidinyl, 4-piperidylbenzoic acid, cambridge id 5267560
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material 4-Piperidinobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.