missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidineacetic acid hydrochloride, 95%, Thermo Scientific™

Product Code. 15431059
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73415-84-6
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD03414187
InChI Key IYDUFJOXYMLYNM-UHFFFAOYSA-N
Synonym 4-piperidineacetic acid hydrochloride, 2-piperidin-4yl acetic acid hydrochloride, 2-piperidin-4-yl acetic acid hydrochloride, 4-piperidineacetic acid hcl, 4-piperidineacetic acid, hydrochloride, piperidin-4-ylacetic acid hydrochloride, piperidine-4-acetic acid hcl, piperidine-4-acetic acid hydrochloride, 4-piperidine acetic acid hydrochloride, piperidin-4-yl-acetic acid hcl
PubChem CID 15565775
IUPAC Name 2-piperidin-4-ylacetic acid;hydrochloride
SMILES C1CNCCC1CC(=O)O.Cl

Specifications

Color Yellow
Formula Weight 179.64
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material 4-Piperidineacetic acid hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'