Learn More
4-(Piperidin-1-ylmethyl)benzonitrile, ≥97%, Maybridge™
Brand: Maybridge CC45516DE
Code : 31
Additional Details : CAS Number : 727733-92-8 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
727733-92-8 | |
200.285 | |
JOGVBKNLRQCYKS-UHFFFAOYSA-N | |
7537603 | |
C1CCN(CC1)CC2=CC=C(C=C2)C#N |
C13H16N2 | |
MFCD06796182 | |
4-piperidin-1-ylmethyl benzonitrile, 1-4-cyanobenzyl piperidine, 4-piperidinomethyl benzonitrile, 4-piperidylmethyl benzenecarbonitrile, 4-piperidin-1-ylmethyl-benzonitrile, benzonitrile,4-1-piperidinylmethyl | |
4-(piperidin-1-ylmethyl)benzonitrile |
Specifications
4-(Piperidin-1-ylmethyl)benzonitrile | |
727733-92-8 | |
100.0 | |
C13H16N2 | |
4-piperidin-1-ylmethyl benzonitrile, 1-4-cyanobenzyl piperidine, 4-piperidinomethyl benzonitrile, 4-piperidylmethyl benzenecarbonitrile, 4-piperidin-1-ylmethyl-benzonitrile, benzonitrile,4-1-piperidinylmethyl | |
C1CCN(CC1)CC2=CC=C(C=C2)C#N | |
200.285 | |
200.28 |
5g | |
96.5 | |
97% | |
MFCD06796182 | |
JOGVBKNLRQCYKS-UHFFFAOYSA-N | |
4-(piperidin-1-ylmethyl)benzonitrile | |
7537603 | |
97% |