missing translation for 'onlineSavingsMsg'
Learn More

4-(piperidin-1-ylmethyl)benzaldehyde, 95%, Thermo Scientific™

Product Code. 10245564
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10245564 5 g 5g
10598053 250 mg 250mg
10128092 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245564 Supplier Thermo Scientific Maybridge Supplier No. CC45504DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 471929-86-9
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
InChI Key WWBOSCKXPMBVLG-UHFFFAOYSA-N
PubChem CID 7164649
IUPAC Name 4-(piperidin-1-ylmethyl)benzaldehyde
SMILES C1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 4-piperidin-1-ylmethyl benzaldehyde, 4-1-piperidinylmethyl benzaldehyde, benzaldehyde,4-1-piperidinylmethyl, 4-piperidylmethyl benzaldehyde, 4-piperidinomethyl benzaldehyde, benzaldehyde, 4-1-piperidinylmethyl
Formula Weight 203.28
Percent Purity 95%
Chemical Name or Material 4-(Piperidin-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.