missing translation for 'onlineSavingsMsg'
Learn More

4-(Piperazin-1-yl)-6-(pyrrolidin-1-yl)pyrimidine, TRC

Product Code. 30428117
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428117 25 mg 25mg
30410910 10 mg -
30416269 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428117 Supplier Toronto Research Chemicals Supplier No. TRCP24635125MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1105193-40-5
Molecular Formula C12H19N5
Molecular Weight (g/mol) 233.31
IUPAC Name 4-piperazin-1-yl-6-pyrrolidin-1-ylpyrimidine
SMILES C1CCN(C1)c2cc(ncn2)N3CCNCC3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H19N5/c1-2-6-16(5-1)11-9-12(15-10-14-11)17-7-3-13-4-8-17/h9-10,13H,1-8H2
Formula Weight 233.164
Chemical Name or Material 4-(Piperazin-1-yl)-6-(pyrrolidin-1-yl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.