missing translation for 'onlineSavingsMsg'
Learn More

4-Picoline, 99%, Thermo Scientific Chemicals

Catalog Number 10317243
12.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
108-89-4
C6H7N
93.13
MFCD00006440
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy
7963
CHEBI:32547
4-methylpyridine
CC1=CC=NC=C1
108-89-4
C6H7N
93.13
MFCD00006440
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy
7963
CHEBI:32547
4-methylpyridine
CC1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 2.4°C
Color Yellow
Density 0.9500g/mL
Boiling Point 145°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.506
Quantity 250 mL
Beilstein 20, 240
Merck Index 15, 7514
Specific Gravity 0.95
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 93.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Picoline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'