missing translation for 'onlineSavingsMsg'
Learn More

4-Phthalimido-2-butyne, 97%, Thermo Scientific™

Product Code. 10519813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10519813 1 g Glass bottle 1g
10310684 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10519813 Supplier Thermo Scientific Acros Supplier No. 382150010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 113439-83-1
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD00160830
InChI Key COPWPECHFWIYSP-UHFFFAOYSA-N
Synonym n-2-butynyl phthalimide, 2-but-2-yn-1-yl isoindoline-1,3-dione, 2-but-2-yn-1-yl isoindole-1,3-dione, acmc-1bscb, 1-phthalimido-2-butyne, 4-phthalimido-2-butyne, 2-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-butyn-1-yl
PubChem CID 8063352
IUPAC Name 2-(but-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES CC#CCN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 217.0°C to 218.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 199.21
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Phthalimido-2-butyne
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phthalimido-2-butyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.