missing translation for 'onlineSavingsMsg'
Learn More

4α-Phorbol 12-Myristate 13-Acetate, MP Biomedicals™

Product Code. 15937285 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 15937285

Brand: MP Biomedicals 0218388205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synonym: Phorbol 12-Myristate 13-Acetate, 12-O-Tetradecanoylphorbol-13-Acetate, Tetradecanoylphorbol Acetate

Negative control for phorbol ester activation of protein kinase C.

TRUSTED_SUSTAINABILITY

Specifications

CAS 63597-44-4
Quantity 5 mg
Molecular Formula C36H56O8
Physical Form Solid
Purity ≥99%
InChI Key PHEDXBVPIONUQT-LQLWEASQSA-N
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
IUPAC Name (1S,2S,6S,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Weight (g/mol) 616.84
compliance-icons
Product Identifier
  • phorbol-1,2-myristate-1,3-acetate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : phorbol-1,2-myistate-1,3-acetate

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.