missing translation for 'onlineSavingsMsg'
Learn More

(4-phenyltetrahydropyran-4-yl)methylamine, 95%, Thermo Scientific™

Product Code. p-4391451
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265474 250 mg Amber Glass Bottle 250mg
10022414 1 g Amber Glass Bottle 1g
10290255 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265474 Supplier Thermo Scientific Maybridge Supplier No. CC69113CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14006-32-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD02946795
InChI Key IPYDNTFWMDEOBW-UHFFFAOYSA-N
Synonym 4-phenyloxan-4-yl methanamine, 4-phenyltetrahydro-2h-pyran-4-yl methanamine, 4-phenyltetrahydropyran-4-yl methylamine, 1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, c-4-phenyl-tetrahydro-pyran-4-yl-methylamine, 4-phenyltetrahydro-2h-pyran-4-yl methylamine, 1-4-phenyloxan-4-yl methanamine, 2h-pyran-4-methanamine, tetrahydro-4-phenyl, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine, 4-phenyl-tetrahydro-2h-pyran-4-methylamine
PubChem CID 6483779
SMILES NCC1(CCOCC1)C1=CC=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-(4-phenyloxan-4-yl)methanamine
Formula Weight 191.27
Percent Purity 95%
Chemical Name or Material (4-phenyltetrahydropyran-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.