missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenylsulfonyl)thiophen-3-amine, ≥95%, Maybridge

Product Code. 10394232
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10394232 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10394232 Supplier Thermo Scientific Maybridge Supplier No. KM01765DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175201-60-2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C10H9NO2S2
MDL Number MFCD00067914
Quantity 1 g
Synonym 4-phenylsulfonyl thiophen-3-amine, 4-benzenesulfonyl thiophen-3-amine, 4-phenylsulfonyl thiophene-3-amine, maybridge1_004632, 4-phenylsulfonyl-3-thienylamine, 4-benzenesulfonyl-3-thiophenamine, 3-thiophenamine,4-phenylsulfonyl
InChI Key HYLCXPMYGZOANR-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)S(=O)(=O)C2=CSC=C2N
IUPAC Name 4-(benzenesulfonyl)thiophen-3-amine
Molecular Weight (g/mol) 239.307
PubChem CID 2778524
Formula Weight 239.32
Percent Purity 95%
Chemical Name or Material 4-(Phenylsulfonyl)thiophen-3-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.