missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Phenylsemicarbazide, 98+%

Product Code. 11364679
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364679 10 g 10g
11457256 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364679 Supplier Alfa Aesar™ Supplier No. B25578.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 537-47-3
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD00007590
InChI Key MOCKWYUCPREFCZ-UHFFFAOYSA-N
Synonym 4-phenylsemicarbazide, n-phenylhydrazinecarboxamide, 3-amino-1-phenylurea, hydrazinecarboxamide, n-phenyl, anilinoformylhydrazine, unii-bpu96sx57a, semicarbazide, 4-phenyl, bpu96sx57a, hydrazinecarboxanilide, anilinocarbonylhydrazine
PubChem CID 10837
IUPAC Name 1-amino-3-phenylurea
SMILES C1=CC=C(C=C1)NC(=O)NN

Specifications

Melting Point 123°C to 125°C
Quantity 10 g
Beilstein 511107
Sensitivity Air Sensitive
Merck Index 14,7312
Formula Weight 151.17
Percent Purity ≥98%
Chemical Name or Material 4-Phenylsemicarbazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.