missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylpiperidine, 96%

Product Code. 15452739
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452739 1 g 1g
15462739 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452739 Supplier Thermo Scientific Alfa Aesar Supplier No. H66389.03
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 771-99-3 | C11H15N | 161.248 g/mol

4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-99-3
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00006002
InChI Key UTBULQCHEUWJNV-UHFFFAOYSA-N
Synonym 4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
PubChem CID 69873
IUPAC Name 4-phenylpiperidine
SMILES C1CNCCC1C2=CC=CC=C2

Specifications

Melting Point 61°C to 64°C
Color Yellow to Brown
Quantity 1 g
Beilstein 124508
Solubility Information Soluble in water.
Formula Weight 161.24
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Phenylpiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.