missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylphenol, 97%

Product Code. 11419762
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419762 500 g 500g
11409762 100 g 100g
11429762 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419762 Supplier Thermo Scientific Alfa Aesar Supplier No. A10817.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 92-69-3 | C12H10O | 170.211 g/mol

Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

Solubility
Soluble in water (0.7 mg/ml at 20°C), ether, most common organic solvents, methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-69-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00002347
InChI Key YXVFYQXJAXKLAK-UHFFFAOYSA-N
Synonym 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
PubChem CID 7103
ChEBI CHEBI:34422
IUPAC Name 4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)O

Specifications

Melting Point 164°C to 168°C
Boiling Point 305°C to 308°C
Flash Point 165°C (329°F)
Quantity 500 g
UN Number UN3077
Beilstein 1907452
Merck Index 14,7305
Solubility Information Soluble in water (0.7mg/ml at 20°C),ether,most common organic solvents,methanol,and chloroform.
Formula Weight 170.21
Percent Purity 98%
Chemical Name or Material 4-Phenylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Hydroxybiphenyl
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (1,1-Biphenyl)-4-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.