missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Phenylethynyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™
£54.30 - £173.00
Chemical Identifiers
CAS | 1190376-20-5 |
---|---|
Molecular Formula | C20H21BO2 |
Molecular Weight (g/mol) | 304.196 |
MDL Number | MFCD16294498 |
InChI Key | AAIDERXZGJSORP-UHFFFAOYSA-N |
Synonym | 4,4,5,5-tetramethyl-2-4-phenylethynyl-phenyl-1,3,2 dioxaborolane, 4-phenylethynyl benzeneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-4-2-phenylethynyl phenyl-1,3,2-dioxaborolane, amtb199, 4-phenylethynyl phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylethynyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-phenylethynyl phenyl-1,3,2-dioxaborolane |
PubChem CID | 57416880 |
IUPAC Name | 4,4,5,5-tetramethyl-2-[4-(2-phenylethynyl)phenyl]-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CC3=CC=CC=C3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419275
|
Thermo Scientific Alfa Aesar
H51699.03 |
1 g |
£54.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15429275
|
Thermo Scientific Alfa Aesar
H51699.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1190376-20-5 | |
304.196 | |
AAIDERXZGJSORP-UHFFFAOYSA-N | |
57416880 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CC3=CC=CC=C3 |
C20H21BO2 | |
MFCD16294498 | |
4,4,5,5-tetramethyl-2-4-phenylethynyl-phenyl-1,3,2 dioxaborolane, 4-phenylethynyl benzeneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-4-2-phenylethynyl phenyl-1,3,2-dioxaborolane, amtb199, 4-phenylethynyl phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylethynyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-phenylethynyl phenyl-1,3,2-dioxaborolane | |
4,4,5,5-tetramethyl-2-[4-(2-phenylethynyl)phenyl]-1,3,2-dioxaborolane |
Specifications
1190376-20-5 | |
MFCD16294498 | |
4,4,5,5-tetramethyl-2-4-phenylethynyl-phenyl-1,3,2 dioxaborolane, 4-phenylethynyl benzeneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-4-2-phenylethynyl phenyl-1,3,2-dioxaborolane, amtb199, 4-phenylethynyl phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylethynyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-phenylethynyl phenyl-1,3,2-dioxaborolane | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CC3=CC=CC=C3 | |
304.196 | |
304.19 | |
4-(Phenylethynyl)benzeneboronic acid pinacol ester |
C20H21BO2 | |
20037873 | |
AAIDERXZGJSORP-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-[4-(2-phenylethynyl)phenyl]-1,3,2-dioxaborolane | |
57416880 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures