missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcyclohexanone, 98+%

Product Code. 11392606
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392606 25 g 25g
11302616 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392606 Supplier Thermo Scientific Alfa Aesar Supplier No. L08614.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4894-75-1 | C12H14O | 174.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4894-75-1
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00001641
InChI Key YKAYMASDSHFOGI-UHFFFAOYSA-N
Synonym 4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
PubChem CID 78605
IUPAC Name 4-phenylcyclohexan-1-one
SMILES C1CC(=O)CCC1C2=CC=CC=C2

Specifications

Melting Point 78°C to 82°C
Boiling Point 158°C to 160°C (12 mmHg)
Flash Point >100°C (212°F)
Quantity 25 g
Beilstein 2045904
Formula Weight 174.24
Percent Purity ≥98%
Chemical Name or Material 4-Phenylcyclohexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.