missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcyclohexanone, 98+%

Product Code. 10197413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10197413 10 g Glass bottle 10g
10164060 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197413 Supplier Thermo Scientific Acros Supplier No. 187940100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4894-75-1 | C12H14O | 174.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4894-75-1
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD00001641
InChI Key YKAYMASDSHFOGI-UHFFFAOYSA-N
Synonym 4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
PubChem CID 78605
IUPAC Name 4-phenylcyclohexan-1-one
SMILES C1CC(=O)CCC1C2=CC=CC=C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 77°C to 80°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H9(=O)
Quantity 10 g
Beilstein 07, III, 1466
Solubility Information Solubility in water: slightly soluble
Formula Weight 174.24
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylcyclohexanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.