missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenylamino)-1-benzyl-4-piperidinecarboxamide, TRC

Product Code. 30425963
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425963 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425963 Supplier Toronto Research Chemicals Supplier No. TRCP30748525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Phenylamino)-1-benzyl-4-piperidinecarboxamide
CAS 1096-03-3
Molecular Formula C19 H23 N3 O
Quantity 25 mg
Synonym 4-Piperidinecarboxamide','4-(phenylamino)-1-(phenylmethyl)-, Isonipecotamide','4-anilino-1-benzyl- (7CI,8CI), 4-(Phenylamino)-1-(phenylmethyl)-4-piperidinecarboxamide, 1-Benzyl-4-(phenylamino)piperidine-4-carboxamide, 4-Anilino-1-benzyl-4-piperidinecarboxamide, NSC 73749, 1-Benzyl-4-(phenylamino)piperidine-4-carboxamide
InChI Formula InChI=1S/C19H23N3O/c20-18(23)19(21-17-9-5-2-6-10-17)11-13-22(14-12-19)15-16-7-3-1-4-8-16/h1-10,21H,11-15H2,(H2,20,23)
SMILES NC(=O)C1(CCN(Cc2ccccc2)CC1)Nc3ccccc3
IUPAC Name 4-anilino-1-benzylpiperidine-4-carboxamide
Molecular Weight (g/mol) 309.41
Formula Weight 309.1841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.