missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-5-(trifluoromethyl)thiophen-3-amine, ≥97%, Thermo Scientific™

Product Code. 10832051
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 256427-77-7
Molecular Formula C11H8F3NS
Molecular Weight (g/mol) 243.247
MDL Number MFCD01934528
InChI Key KBIPYEXWOYDDDL-UHFFFAOYSA-N
Synonym 4-phenyl-5-trifluoromethyl thiophen-3-amine, 4-phenyl-5-trifluoromethyl thien-3-ylamine, 3-amino-4-phenyl-5-trifluoromethyl thiophene, maybridge3_006438, 4-phenyl-5-trifluoromethyl-3-thienylamine, 4-phenyl-5-trifluoromethyl thiophene-3-amine
PubChem CID 2782905
IUPAC Name 4-phenyl-5-(trifluoromethyl)thiophen-3-amine
SMILES C1=CC=C(C=C1)C2=C(SC=C2N)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 243.25
Percent Purity 97%
Chemical Name or Material 4-Phenyl-5-(trifluoromethyl)thiophen-3-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'