missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-butyn-2-one, 98%, Thermo Scientific Chemicals

Catalog Number p-7077176
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1817-57-8
C10H8O
144.17
UPEUQDJSUFHFQP-UHFFFAOYSA-N
4-phenyl-3-butyn-2-one, 3-butyn-2-one, 4-phenyl, 1-phenyl-1-butyn-3-one, phenylbutynone, acetylphenyl-acetylene, acmc-1boyr, 4-phenyl-3-butyn-2-on, 4-phenyl-3-batyu-2-one, 4-phenyl-3-butyn-2-0ne, 4-phenyl-but-3-yn-2-one
74555
CHEBI:51731
4-phenylbut-3-yn-2-one
CC(=O)C#CC1=CC=CC=C1
This item is not returnable. View return policy
1817-57-8
C10H8O
144.17
UPEUQDJSUFHFQP-UHFFFAOYSA-N
4-phenyl-3-butyn-2-one, 3-butyn-2-one, 4-phenyl, 1-phenyl-1-butyn-3-one, phenylbutynone, acetylphenyl-acetylene, acmc-1boyr, 4-phenyl-3-butyn-2-on, 4-phenyl-3-batyu-2-one, 4-phenyl-3-butyn-2-0ne, 4-phenyl-but-3-yn-2-one
74555
CHEBI:51731
4-phenylbut-3-yn-2-one
CC(=O)C#CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9900g/mL
Boiling Point 75°C to 76°C (1.1 hPa)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5C≡CCOCH3
Refractive Index 1.5762
Quantity 1 g
Beilstein 07,385
Specific Gravity 0.99
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material 4-Phenyl-3-butyn-2-one
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenyl-3-butyn-2-one, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'