missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-buten-2-one, 98+%, Thermo Scientific Chemicals

Product Code. 10519993
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-57-6
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00008779
InChI Key BWHOZHOGCMHOBV-BQYQJAHWSA-N
Synonym benzalacetone, benzylideneacetone, methyl styryl ketone, 4-phenylbut-3-en-2-one, acetocinnamone, 4-phenyl-3-buten-2-one, e-4-phenylbut-3-en-2-one, benzylidene acetone, trans-benzalacetone, benzalaceton
PubChem CID 637759
ChEBI CHEBI:78399
IUPAC Name (E)-4-phenylbut-3-en-2-one
SMILES CC(=O)\C=C\C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 38.5°C
Color Yellow
Boiling Point 261°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCOCH3
Quantity 1 kg
Merck Index 15, 1140
Solubility Information Solubility in water: 1.3g/L (20°C). Other solubilities: sparingly soluble in petroleum ether,freely soluble in alcohol,benzene,chloroform,,ether
Formula Weight 146.19
Percent Purity 98+%
Physical Form Low Melting Solid
Chemical Name or Material 4-Phenyl-3-buten-2-one
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenyl-3-buten-2-one
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'