missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-butyne, 98%

Product Code. p-7046054
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11387806

Brand: Thermo Scientific Alfa Aesar L12946.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16520-62-0 | C10H10 | 130.19 g/mol

4-Phenyl-1-butyne is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-butyne is used as an intermediate in chemical research.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16520-62-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00041667
InChI Key QDEOKXOYHYUKMS-UHFFFAOYSA-N
Synonym 4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne
PubChem CID 123360
IUPAC Name but-3-ynylbenzene
SMILES C#CCCC1=CC=CC=C1

Specifications

Density 0.926
Boiling Point 189°C to 190°C
Linear Formula C6H5(CH2)2C≡CH
Refractive Index 1.521
Quantity 5 g
Beilstein 1362221
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 4-Phenyl-1-butyne

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.