missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-butanol, 97%

Product Code. 10164340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164340 50 g Glass bottle 50g
10488920 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10164340 Supplier Thermo Scientific Acros Supplier No. 180370500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3360-41-6 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3360-41-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002971
InChI Key LDZLXQFDGRCELX-UHFFFAOYSA-N
Synonym 4-phenyl-1-butanol, benzenebutanol, 4-phenylbutanol, phenylbutyl alcohol, 4-phenylbutyl alcohol, 1-butanol, 4-phenyl, 4-phenyl butanol-1, 4-phenyl butanol, unii-q5orz1321g, 4-phenyl-n-butanol
PubChem CID 76889
IUPAC Name 4-phenylbutan-1-ol
SMILES C1=CC=C(C=C1)CCCCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9750g/mL
Boiling Point 140°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)4OH
Refractive Index 1.5200 to 1.522
Quantity 50 g
Beilstein 06, II, 487
Specific Gravity 0.975
Formula Weight 150.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Phenyl-1-butanol, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.