missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1,2,3-thiadiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10160192
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160192 1 g Amber Glass Bottle 1g
10611483 250 mg Amber Glass Bottle 250mg
10420954 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10160192 Supplier Thermo Scientific Maybridge Supplier No. CC21501DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 78875-63-5
Molecular Formula C9H6N2O2S
Molecular Weight (g/mol) 206.219
MDL Number MFCD00052216
InChI Key DQZLGXDVESJWKA-UHFFFAOYSA-N
Synonym 4-phenyl-1,2,3-thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-phenyl, 4-phenyl-5-thiadiazolecarboxylic acid, 5-carboxy-4-phenyl-1,2,3-thiadiazole, 4-phenyl-1,2,3-thiadiazol-5-carboxylic acid, 4-phenyl 1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylicacid, 4-phenyl, 4-phenyl-1,2,3 thiadiazole-5-carboxylic acid
PubChem CID 2737167
IUPAC Name 4-phenylthiadiazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SN=N2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1, 2, 3-thiadiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.