missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxypiperidine, 98%

Product Code. 15417907
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417907 5 g 5g
11479750 250 mg 250mg
11489750 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15417907 Supplier Thermo Scientific Alfa Aesar Supplier No. H50984.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3202-33-3 | C11H15NO | 177.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3202-33-3
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD04114971
InChI Key KBYPITRKIJKGMD-UHFFFAOYSA-N
Synonym piperidine, 4-phenoxy, unii-aof4m4l61z, 4-phenoxy-piperidine, aof4m4l61z, piperidine, 4-phenoxy-, hydrochloride, 4-phenoxy piperidine, acmc-209hq3
PubChem CID 18878
IUPAC Name 4-phenoxypiperidine
SMILES C1CC(CCN1)OC1=CC=CC=C1

Specifications

Boiling Point 107°C to 108°C (0.1 mmHg)
Quantity 5 g
Formula Weight 177.25
Percent Purity 99%
Chemical Name or Material 4-Phenoxypiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.