missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyphenylmagnesium bromide, 0.5 M in 2-MeTHF

Product Code. 15471156
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471156 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15471156 Supplier Thermo Scientific Alfa Aesar Supplier No. H54411.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21473-02-9 | C12H9BrMgO | 273.41 g/mol

4-Phenoxyphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenoxyphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and base
TRUSTED_SUSTAINABILITY

Specifications

CAS 21473-02-9
Molecular Formula C12H9BrMgO
MDL Number MFCD00672009
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 4-phenoxyphenylmagnesium bromide, p-phenoxyphenylmagnesium bromide, p-phenoxyphenyl magnesium bromide, 4-phenoxy-phenylmagnesium bromide, 4-phenoxyphenyl magnesium bromide, dtikyzptjrzftl-uhfffaoysa-m, magnesium,bromo 4-phenoxyphenyl, bromo 4-phenoxyphenyl magnesium, 4-phenoxyphenylmagnesium bromide solution, 4-phenoxyphenylmagnesium bromide, 0.5 m in 2-methf, grignard reagent
Solubility Information It reacts with water.
InChI Key DTIKYZPTJRZFTL-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC=C(OC2=CC=CC=C2)C=C1
IUPAC Name magnesium;phenoxybenzene;bromide
Molecular Weight (g/mol) 273.41
PubChem CID 13467796
Formula Weight 273.41
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 4-Phenoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.