missing translation for 'onlineSavingsMsg'
Learn More

(4-Phenoxyphenyl)methylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10230035
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10230035 5 g Amber Glass Bottle 5g
10776694 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10230035 Supplier Thermo Scientific Maybridge Supplier No. CC53763DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 169944-04-1
Molecular Formula C13H14ClNO
Molecular Weight (g/mol) 235.711
InChI Key VHCSCKHIGGFTHN-UHFFFAOYSA-N
Synonym 4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine hydrochloride, 4-phenoxybenzylamine hydrochloride, benzenemethanamine, 4-phenoxy-, hydrochloride, 1-4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine, chloride, 1-4-phenoxyphenyl methanamine-hydrogen chloride 1/1
PubChem CID 22293026
IUPAC Name (4-phenoxyphenyl)methanamine;hydrochloride
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CN.Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 235.71
Percent Purity 97%
Chemical Name or Material (4-Phenoxyphenyl)methylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.