missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyphenyl isocyanate, 98%

Product Code. 11392567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392567 1 g 1g
11302577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392567 Supplier Thermo Scientific Alfa Aesar Supplier No. L11686.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59377-19-4 | C13H9NO2 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59377-19-4
Molecular Formula C13H9NO2
Molecular Weight (g/mol) 211.22
MDL Number MFCD00013876
InChI Key PNBUGOFIKAHZRW-UHFFFAOYSA-N
Synonym 4-phenoxyphenyl isocyanate, 4-phenoxyphenylisocyanate, 1-isocyanato-4-phenoxy-benzene, benzene, 1-isocyanato-4-phenoxy, acmc-20ambk, pubchem17221, p-phenoxyphenylisocyanate, 4-phenoxybenzenisocyanate, 4-phenoxyphenyl-isocyanate, 4-isocyanatodiphenyl ether
PubChem CID 2734896
IUPAC Name 1-isocyanato-4-phenoxybenzene
SMILES O=C=NC1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Density 1.169
Boiling Point 169°C to 170°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 1 g
UN Number UN2206
Beilstein 393196
Sensitivity Moisture sensitive
Formula Weight 211.22
Percent Purity 98%
Chemical Name or Material 4-Phenoxyphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.