missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyphenethylamine, 97+%, Thermo Scientific™

Product Code. 10208383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10208383 1 g Glass bottle 1g
10617274 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10208383 Supplier Thermo Scientific Acros Supplier No. 393490010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 118468-18-1
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00079777
InChI Key JNHPLGDXCJAUBX-UHFFFAOYSA-N
PubChem CID 145535
IUPAC Name 2-(4-phenoxyphenyl)ethanamine
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CCN

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Yellow
Density 1.0900g/mL
Boiling Point 125°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OC6H4CH2CH2NH2
Refractive Index 1.5830 to 1.5850 (20°C, 589nm)
Quantity 1 g
Beilstein 13, IV, 1789
Specific Gravity 1.09
Synonym 4-phenoxyphenethylamine, 2-4-phenoxyphenyl ethanamine, 2-4-phenoxyphenyl ethan-1-amine, o-phenyl-tyramine, benzeneethanamine,4-phenoxy, benzeneethanamine, 4-phenoxy, 2-4-phenoxy-phenyl-ethylamine, 4-butylpiperidine hcl form, acmc-1cb59, 2-4-phenoxyphenyl ethylamine
Formula Weight 213.28
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 4-Phenoxyphenethylamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenoxyphenethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.