missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenoxybenzylamine, MedChemExpress
Description
4-Phenoxybenzylamine inhibits the function of the NS3 protein by stabilizing an inactive conformation with an IC50 of about 500 μM against FL NS3/4a.
Specifications
Specifications
| Chemical Name or Material | 4-Phenoxybenzylamine |
| CAS | 107622-80-0 |
| Color | Colorless |
| Purity Grade Notes | Research |
| Molecular Formula | C13H13NO |
| Quantity | 100 mg |
| Solubility Information | DMSO : 12.5 mg/mL (62.74 mM; Need ultrasonic) |
| SMILES | NCC1=CC=C(OC2=CC=CC=C2)C=C1 |
| Molecular Weight (g/mol) | 199.25 |
| Formula Weight | 199.25 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?