missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzeneboronic acid, 98%

Product Code. 15449717
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449717 5 g 5g
15439717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449717 Supplier Thermo Scientific Alfa Aesar Supplier No. H52950.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51067-38-0 | C12H11BO3 | 214.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51067-38-0
Molecular Formula C12H11BO3
Molecular Weight (g/mol) 214.03
MDL Number MFCD00093312
InChI Key KFXUHRXGLWUOJT-UHFFFAOYSA-N
Synonym 4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid, pubchem7890, acmc-209krn, phenoxyphenylboronic acid, d0d2ja
PubChem CID 2734377
IUPAC Name (4-phenoxyphenyl)boronic acid
SMILES OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 141°C to 145°C
Quantity 5 g
Formula Weight 214.03
Percent Purity 98%
Chemical Name or Material 4-Phenoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.