missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyaniline, 97%

Product Code. 10306920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306920 100 g Glass bottle 100g
10560332 5 g Glass bottle 5g
10167553 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306920 Supplier Thermo Scientific Acros Supplier No. 130231000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 139-59-3 | C12H11NO | 185.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-59-3
MDL Number MFCD00007862
InChI Key WOYZXEVUWXQVNV-UHFFFAOYSA-N
Synonym p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether
PubChem CID 8764
IUPAC Name 4-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 82°C to 86°C
Color Brown to Brown-Green
Boiling Point 140°C (3.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H11NO
Linear Formula C6H5OC6H4NH2
Quantity 100 g
Beilstein 13, 438
Solubility Information Solubility in water: < 1g/l in water (20°C). Other solubilities: soluble in isopropanol,toluene,very soluble in acetone,1,2-dichloroethane
Molecular Weight (g/mol) 185.23
Formula Weight 185.23
Percent Purity 97%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 4-Phenoxyaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Phenoxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.