missing translation for 'onlineSavingsMsg'
Learn More

4'-Phenoxyacetophenone, 98%, ACROS Organics™

Product Code. 10011551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10011551 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10011551 Supplier Thermo Scientific Acros Supplier No. 334980250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5031-78-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color Yellow
Boiling Point 200.0°C (12.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C14H12O2
MDL Number MFCD00008744
Quantity 25 g
Synonym 4'-phenoxyacetophenone, 1-4-phenoxyphenyl ethanone, 4-phenoxyacetophenone, p-phenoxyacetophenone, 1-4-phenoxyphenyl ethan-1-one, ethanone, 1-4-phenoxyphenyl, 4-acetyldiphenyl ether, 1-4-phenoxyphenyl-1-ethanone, 1-4-phenoxyphenyl ethanon, pubchem13195
Solubility Information Solubility in water: insoluble.
InChI Key DJNIFZYQFLFGDT-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
IUPAC Name 1-(4-phenoxyphenyl)ethan-1-one
Molecular Weight (g/mol) 212.25
PubChem CID 236783
Formula Weight 212.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4'-Phenoxyacetophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Phenoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.