missing translation for 'onlineSavingsMsg'
Learn More

4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, Thermo Scientific Chemicals

Product Code. 10239403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10239403 5 g 5g
10133700 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10239403 Supplier Thermo Scientific Acros Supplier No. 123980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 10580-19-5 , 825-90-1
Molecular Formula C6H5O4S
Molecular Weight (g/mol) 173.16
MDL Number MFCD00044734,MFCD00150724
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
PubChem CID 23666329
SMILES OC1=CC=C(C=C1)S([O-])(=O)=O

Specifikationer

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color Green to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H4SO3Na·2H2O
Quantity 5 g
Solubility Information Solubility in water: 24g/100mL.
IUPAC Name sodium;4-hydroxybenzenesulfonate;dihydrate
Formula Weight 232.18
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Phenolsulfonic acid, sodium salt dihydrate, 99%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 4-Phenolsulfonic acid, sodium salt dihydrate
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.