missing translation for 'onlineSavingsMsg'
Learn More

4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, Thermo Scientific Chemicals

Product Code. 10133700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10133700 100 g 100g
10239403 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133700 Supplier Thermo Scientific Acros Supplier No. 123981000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10580-19-5 , 825-90-1
Molecular Formula C6H5O4S
Molecular Weight (g/mol) 173.16
MDL Number MFCD00044734,MFCD00150724
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
PubChem CID 23666329
SMILES OC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color Green to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula HOC6H4SO3Na·2H2O
Quantity 100 g
Solubility Information Solubility in water: 24g/100mL.
IUPAC Name sodium;4-hydroxybenzenesulfonate;dihydrate
Formula Weight 232.18
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Phenolsulfonic acid, sodium salt dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenolsulfonic acid, sodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.