missing translation for 'onlineSavingsMsg'
Learn More

4-Pentynoic acid, 95%

Product Code. 11347326
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347326 1 g 1g
11357326 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347326 Supplier Thermo Scientific Alfa Aesar Supplier No. L09282.03
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6089-09-4 | C5H5O2 | 97.09 g/mol

4-Pentynoic acid is widely utilized as a building block for the synthesis of eight sequence-defined model oligomers. Due to its triple bond at terminal carbon, it is employed as an intermediate to produce biologically active compounds. It is used as a hypoglycemic agent, which increases liver tyrosine aminotransferase and plasma corticosterone while decreasing blood sugar in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentynoic acid is widely utilized as a building block for the synthesis of eight sequence-defined model oligomers. Due to its triple bond at terminal carbon, it is employed as an intermediate to produce biologically active compounds. It is used as a hypoglycemic agent, which increases liver tyrosine aminotransferase and plasma corticosterone while decreasing blood sugar in rats.

Solubility
Soluble in low polarity organic solvents. Soluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6089-09-4
MDL Number MFCD00004407

Specifications

Melting Point 51°C to 58°C
Boiling Point 109°C to 110°C (30 mmHg)
Flash Point 75°C (167°F)
Molecular Formula C5H5O2
Linear Formula HC≡C(CH2)2CO2H
Quantity 1 g
UN Number UN3261
Beilstein 1742047
Sensitivity Air and light sensitive; Hygroscopic
Synonym 4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d
Solubility Information Soluble in low polarity organic solvents. Soluble in water.
InChI Key MLBYLEUJXUBIJJ-UHFFFAOYSA-M
SMILES C#CCCC(=O)O
IUPAC Name pent-4-ynoic acid
Molecular Weight (g/mol) 97.09
PubChem CID 22464
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material 4-Pentynoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.