missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-1-ol, TRC

Product Code. 30422314
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422314 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422314 Supplier Toronto Research Chemicals Supplier No. TRCP22754025G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Pentyn-1-ol
CAS 5390-04-5
Molecular Formula C5H8O
Quantity 25 g
Synonym (3-Hydroxypropyl)acetylene, 1-Hydroxy-4-pentyne, 1-Pentyn-5-ol, 5-Hydroxy-1-pentyne, NSC 5274, Pent-4-yne-1-ol
InChI Formula InChI=1S/C5H8O/c1-2-3-4-5-6/h1,6H,3-5H2
SMILES OCCCC#C
IUPAC Name pent-4-yn-1-ol
Molecular Weight (g/mol) 84.12
Formula Weight 84.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.