missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-1-ol, 95%

Product Code. 11488593
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488593 100 g 100g
11468593 5 g 5g
11478593 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488593 Supplier Thermo Scientific Alfa Aesar Supplier No. A10405.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5390-04-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Pentyn-1-ol is widely utilized in ring-opening polymerization reactions. It plays an important role as a starting reagent in stereo selective total synthesis of antimicrobial marine metabolites, ieodomycin A and B. It is also used in the preparation of 3-pent-4-ynyloxy phthalonitrile.

Solubility
Miscible with water.

Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5390-04-5
MDL Number MFCD00002974

Specifications

Density 0.907
Boiling Point 153°C to 155°C
Flash Point 62°C (143°F)
Molecular Formula C5H8O
Linear Formula HC≡C(CH2)3OH
Refractive Index 1.445
Quantity 100 g
Beilstein 1736712
Synonym 4-pentyn-1-ol, 4-pentyne-1-ol, 1-pentyn-5-ol, 5-hydroxypentyne, 4-pentynol, 4-pentin-1-ol, pent-4yn-1-ol, 5-hydroxy-1-pentyne, pent-1-yn-5-ol, pubchem2025
Solubility Information Miscible with water.
InChI Key CRWVOXFUXPYTRK-UHFFFAOYSA-N
SMILES C#CCCCO
IUPAC Name pent-4-yn-1-ol
Molecular Weight (g/mol) 84.12
PubChem CID 79346
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 4-Pentyn-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.