missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylphenol, TRC

Product Code. 30132717
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132717 10 g 10g
30132609 5 g 5g
30007652 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132717 Supplier Toronto Research Chemicals Supplier No. TRCP28387510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14938-35-3
Molecular Formula C11 H16 O
Molecular Weight (g/mol) 164.24
Synonym Phenol, p-pentyl- (6CI,7CI,8CI), 4-Pentylphenol, 4-Amylphenol, 4-n-Amylphenol, 4-n-Pentylphenol, Amyl p-hydroxybenzene, p-Amylphenol, p-Hydroxy-n-pentylbenzene, p-Pentylphenol
IUPAC Name 4-pentylphenol
SMILES CCCCCc1ccc(O)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H16O/c1-2-3-4-5-10-6-8-11(12)9-7-10/h6-9,12H,2-5H2,1H3
Formula Weight 164.1201
Chemical Name or Material 4-n-Amylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.