missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzonitrile, 97%, Thermo Scientific™

Product Code. 10191242
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10270-29-8
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD00218832
InChI Key LGPQFJXPPKNHJU-UHFFFAOYSA-N
Synonym 4-n-pentylbenzonitrile, 4-pentylbenzenecarbonitrile, p-amylbenzonitrile, p-n-pentylbenzonitrile, benzonitrile, 4-pentyl
PubChem CID 264951
IUPAC Name 4-pentylbenzonitrile
SMILES CCCCCC1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)4C6H4CN
Quantity 1 g
Formula Weight 173.26
Percent Purity 97%
Chemical Name or Material 4-Pentylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'