missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10673605
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10673605 25 g 25g
10522632 1 g 1g
10029161 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10673605 Supplier Thermo Scientific Maybridge Supplier No. BTB09810EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 73948-18-2
Molecular Formula C11H15ClO2S
Molecular Weight (g/mol) 246.75
MDL Number MFCD00173759
InChI Key XDWXZRVWJHEWMT-UHFFFAOYSA-N
Synonym 4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride
PubChem CID 144625
SMILES CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)4C6H4SO2Cl
Quantity 25 g
IUPAC Name 4-pentylbenzenesulfonyl chloride
Formula Weight 246.76
Percent Purity 97%
Chemical Name or Material 4-Pentylbenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.