missing translation for 'onlineSavingsMsg'
Learn More

(4-Pentenyl)triphenylphosphonium bromide, 97%, Thermo Scientific™

Product Code. 11391447
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11391447

Brand: Thermo Scientific Alfa Aesar L01950.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56771-29-0
Molecular Formula C23H24BrP
Molecular Weight (g/mol) 411.32
MDL Number MFCD00050215
InChI Key KANICXDUFCDRDS-UHFFFAOYSA-M
Synonym 4-pentenyl triphenylphosphonium bromide, pent-4-enyl triphenyl phosphonium bromide, pent-4-en-1-yltriphenylphosphonium bromide, pent-4-en-1-yltriphenylphosphanium bromide, acmc-1az5e, pent-4-enyl triphenyl phosphanium bromide, 4-?pentenyl ?triphenylphosphonium bromide, phosphonium,4-penten-1-yltriphenyl-, bromide 1:1
PubChem CID 10093022
IUPAC Name pent-4-enyl(triphenyl)phosphanium;bromide
SMILES [Br-].C=CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 194°C to 196°C
Quantity 1 g
Beilstein 4171464
Sensitivity Hygroscopic
Formula Weight 411.33
Percent Purity 97%
Chemical Name or Material (4-Pentenyl)triphenylphosphonium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.