missing translation for 'onlineSavingsMsg'
Learn More

4-Pentenoic acid, 99%

Product Code. 10466383 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10466383

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-80-0 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-80-0
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00004408
InChI Key HVAMZGADVCBITI-UHFFFAOYSA-N
Synonym 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
PubChem CID 61138
ChEBI CHEBI:35936
IUPAC Name pent-4-enoic acid
SMILES OC(=O)CCC=C

Specifications

Melting Point -22.5°C
Color Colorless to Yellow
Density 0.9750g/mL
Boiling Point 83.0°C to 84.0°C (12.0 mmHg)
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2CH2CO2H
Refractive Index 1.4273 to 1.4303
Quantity 5 g
Beilstein 02,425
Specific Gravity 0.975
Solubility Information Solubility in water: insoluble
Formula Weight 100.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Pentenoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Pentenoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Pentenoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.