missing translation for 'onlineSavingsMsg'
Learn More

4-Pentenal, 96%

Product Code. 11338066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338066 1 g 1g
11348066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338066 Supplier Thermo Scientific Alfa Aesar Supplier No. L13351.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2100-17-6 | C5H8O | 84.12 g/mol

4-Pentenal is used in the preparation of 6-octen-3-one by addition with ethylene using 2,4-pentanedionatobis(ethylene)rhodium(I) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentenal is used in the preparation of 6-octen-3-one by addition with ethylene using 2,4-pentanedionatobis(ethylene)rhodium(I) as a catalyst.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2100-17-6
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00151841
InChI Key QUMSUJWRUHPEEJ-UHFFFAOYSA-N
Synonym 4-pentenal, 4-penten-1-al, acmc-1cdp8, qumsujwruhpeej-uhfffaoysa
PubChem CID 16418
IUPAC Name pent-4-enal
SMILES C=CCCC=O

Specifications

Density 0.852
Boiling Point 101°C to 105°C
Flash Point 7°C (44°F)
Linear Formula H2C=CH(CH2)2CHO
Refractive Index 1.417
Quantity 1 g
UN Number UN1989
Beilstein 1734369
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material 4-Pentenal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Pentenal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Pentenal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.