missing translation for 'onlineSavingsMsg'
Learn More

4-Penten-1-ol, 99%

Product Code. 10001630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10001630 50 g Glass bottle 50g
10665772 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10001630 Supplier Thermo Scientific Acros Supplier No. 150010500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 821-09-0 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-09-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002975
InChI Key LQAVWYMTUMSFBE-UHFFFAOYSA-N
Synonym 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
PubChem CID 13181
IUPAC Name pent-4-en-1-ol
SMILES C=CCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8340g/mL
Boiling Point 134°C to 137°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CH(CH2)3OH
Refractive Index 1.4289 to 1.4309
Quantity 50 g
Beilstein 01, 443
Specific Gravity 0.834
Solubility Information Solubility in water: 57g/L (25°C)
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Penten-1-ol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Penten-1-ol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.