missing translation for 'onlineSavingsMsg'
Learn More

4-Penten-1-ol, 98+%

Product Code. 11479113
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479113 100 g 100g
11459113 5 g 5g
11469113 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479113 Supplier Thermo Scientific Alfa Aesar Supplier No. A11178.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 821-09-0 | C5H10O | 86.134 g/mol

4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.

Solubility
Solubility in water: 57 g/L (25°C)

Notes
Protect against electrostatic charges. Store away from oxidizing agents. Use only in a chemical fume hood. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-09-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00002975
InChI Key LQAVWYMTUMSFBE-UHFFFAOYSA-N
Synonym 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
PubChem CID 13181
IUPAC Name pent-4-en-1-ol
SMILES C=CCCCO

Specifications

Density 0.84
Boiling Point 134°C to 137°C
Flash Point 43°C (109°F)
Refractive Index 1.43
Quantity 100 g
UN Number UN1987
Beilstein 1560163
Formula Weight 86.13
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 4-Penten-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.